Order Entry
Austria
ContactUsLinkComponent
 
Health
Toxic
Mercury(II) nitrate 0.05 M Hg(NO3)2 (0.1N) ≥99.5% volumetric solution, for chloride determination, Fluka™
Catalog # 34292-1L
CAS Number:  
Health
Toxic
Mercury(II) nitrate 0.05 M Hg(NO3)2 (0.1N) ≥99.5% volumetric solution, for chloride determination, Fluka™
Catalog # 34292-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.995 - 1.005

Specification Test Results

About this item

 :
 :324.60 g/mol
Storage Temperature:Ambient
UN:2024
ADR:6.1,III
 :GHS08
 :GHS06
CAS Number:10045-94-0
Formula:HgN₂O₆
MDL Number:MFCD00011038