


Mercury(II) chloride ≥99.5% ACS, Fluka™
Catalog # 215465-500G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay:Min. 99.5 %
- Residue on ignition a. reducing with HCOOH (SO₄)*:Max. 0.02 %
- Iron (Fe):Max. 0.002 %
- In ether insoluble matter:complying
Specification Test Results
About this item
:
:271.5 g/mol
Merck Index:13,05901
Boiling Pt:302 °C
Melting Pt:276 °C
Density:5.44 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-299-8
UN:1624
ADR:6.1,II
:GHS05
:GHS08
:GHS06
CAS Number:7487-94-7
Formula:HgCl₂
MDL Number:MFCD00011041