Order Entry
Austria
ContactUsLinkComponent
 
Magnesium sulphate heptahydrate ≥99.0% (by complexometric analysis), Puriss. p.a. ACS, Fluka™
 
Magnesium sulphate heptahydrate ≥99.0% (by complexometric analysis), Puriss. p.a. ACS, Fluka™
 

 

  • Appearance:
    complying
  • Assay:
    99.0 - 101.0 %
  • pH (5 %, 25 °C):
    5.0 - 8.2
  • Water insoluble matter:
    Max. 0.005 %
  • Arsenic (As):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.02 %
  • Cadmium (Cd):
    Max. 0.0005 %
  • Cobalt (Co):
    Max. 0.0005 %
  • Chromium (Cr):
    Max. 0.0005 %
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 0.0001 %
  • Potassium (K):
    Max. 0.001 %
  • Manganese (Mn):
    Max. 0.0005 %
  • Sodium (Na):
    Max. 0.001 %
  • Nickel (Ni):
    Max. 0.0005 %
  • Lead (Pb):
    Max. 0.0005 %
  • Strontium (Sr):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 0.0005 %
  • Ammonium (NH₄):
    Max. 0.002 %
  • Chloride (Cl):
    Max. 0.0005 %
  • Nitrate (NO₃):
    Max. 0.002 %
  • Colour of the solution:
    complying
  • Turbidity of solution:
    complying

 

 

Frequently Bought Together