Order Entry
Austria
ContactUsLinkComponent
 
Magnesium sulphate, anhydrous ≥98.0% (by complexometric analysis), extra fine powder, Puriss. p.a. drying agent, Fluka™
 
undefined
Magnesium sulphate, anhydrous ≥98.0% (by complexometric analysis), extra fine powder, Puriss. p.a. drying agent, Fluka™

 

  • Form:
    powder (very fine)
  • Assay:
    98.0 - 102.0 %
  • Loss on ignition (600°C):
    Max. 2 %
  • Aluminium (Al):
    Max. 20 ppm
  • Arsenic (As):
    Max. 0.1 ppm
  • Barium (Ba):
    Max. 5 ppm
  • Bismuth (Bi):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 400 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Potassium (K):
    Max. 200 ppm
  • Lithium (Li):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 20 ppm
  • Molybdenum (Mo):
    Max. 5 ppm
  • Sodium (Na):
    Max. 100 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Strontium (Sr):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 50 ppm
  • Nitrate (NO₃):
    Max. 40 ppm
  • Ammonium (NH₄):
    Max. 20 ppm

 

 

Frequently Bought Together