Order Entry
Austria
ContactUsLinkComponent
Zinc acetate dihydrate 99-102%, Puriss., meets analytical specification of DAB, E650, Fluka™
Zinc acetate dihydrate 99-102%, Puriss., meets analytical specification of DAB, E650, Fluka™
Catalog # 25044-1KG
CAS Number:  
Zinc acetate dihydrate 99-102%, Puriss., meets analytical specification of DAB, E650, Fluka™
Catalog # 25044-1KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    99.0 - 102.0 %
  • pH (5 %, 25 °C):
    6.0 - 8.0
  • Water insoluble matter:
    Max. 0.005 %
  • in CH₃COOH insoluble matters:
    Max. 0.005 %
  • Alkalis/earth alkalis:
    Max. 0.2 %
  • Arsenic (As):
    Max. 3 ppm
  • Calcium (Ca):
    Max. 0.005 %
  • Cadmium (Cd):
    Max. 5 ppm
  • Iron (Fe):
    Max. 50 ppm
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Sodium (Na):
    Max. 0.005 %
  • Lead (Pb):
    Max. 20 ppm
  • Chloride (Cl):
    Max. 50 ppm
  • Sulfate (SO₄):
    Max. 100 ppm
  • Organic volatile impurities (GC):
    complying
  • Residual solvents:
    complying

Specification Test Results

About this item

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate
 :
 :219.51 g/mol
Melting Pt:100 °C
EINECS:209-170-2
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:5970-45-6
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961
Density:1.750 g/cm³ (20 °C)
Merck Index:12,10256