Order Entry
Austria
ContactUsLinkComponent
 
 
 
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
 
undefined
 
 
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
 

 

  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99.90 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.03 %
  • Absorbance at 270 nm:
    Max. 1.0
  • Absorbance 280 nm:
    Max. 0.30
  • Absorbance at 290 nm:
    Max. 0.15
  • Absorbance at 310 nm:
    Max. 0.05
  • Absorbance at 320 nm:
    Max. 0.03
  • Absorbance at 360 nm:
    Max. 0.01
  • Absorbance at 400 nm:
    Max. 0.01
  • Explosion Limit:
    1.8 %, 100 °F
  • Vapor Density:
    3 (vs air)

 

 

Frequently Bought Together