Order Entry
Austria
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP) 99%, Reagent Grade
N-Methyl-2-pyrrolidone (NMP) 99%, Reagent Grade
Certificates
undefined
N-Methyl-2-pyrrolidone (NMP) 99%, Reagent Grade

Specification Test Results

  • Vapor Pressure:
    0.29 mmHg ( 20 °C)
  • IR spectrum:
    complying
  • Assay (GC):
    Min. 98.5 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • APHA:
    Max. 50
  • Vapor Density:
    3.4 (vs air)
  • Explosion Limit:
    9.5 %
  • Autoignition Temperature:
    518 °F
  • Refractive Index:
    n20/D 1.47 (lit.)

Specification Test Results

Related Information

Frequently Bought Together