Order Entry
Austria
ContactUsLinkComponent
 
N-Methyl-2-pyrrolidone (NMP) ≥99%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
 
undefined
N-Methyl-2-pyrrolidone (NMP) ≥99%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

 

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.0 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 350 nm:
    Max. 0.01
  • Absorbance at 325 nm:
    Max. 0.10
  • Absorbance at 300 nm:
    Max. 0.50
  • Absorbance at 285 nm:
    Max. 1.0
  • APHA:
    Max. 10
  • Vapor Density:
    3.4 (vs air)
  • Vapor Pressure:
    0.99 mmHg ( 40 °C)
  • Refractive Index:
    n20/D 1.47 (lit.)
  • Autoignition Temperature:
    518 °F

 

 

Frequently Bought Together