Ammonium sulphamate ≥99.0% (by titrimetric analysis), Puriss. p.a., Fluka™
Catalog # 09960-100G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.0 - 101.0 %
- Melting point:131.5 - 133.5 °C
- pH (5 %, 20°C):5.0 - 6.0
- Colour of the solution:complying
- Clearness of the solution:complying
- Sulfated ash:Max. 0.02 %
- Loss on drying (110°C):Max. 1 %
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 50 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 50 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Chloride (Cl):Max. 10 ppm
- Sulfate (SO₄):Max. 200 ppm
Specification Test Results
About this item
Synonyms: Ammonium sulphamidate, Ammonium amidosulphonate
:
:114.13 g/mol
Melting Pt:131…135 °C
EINECS:231-871-7
:GHS07
Storage Temperature:Ambient
CAS Number:7773-06-0
Formula:H₆N₂O₃S
MDL Number:MFCD00011429
Density:1.00 g/cm³
Merck Index:12,00589