Order Entry
Austria
ContactUsLinkComponent
Potassium dihydrogen phosphate ≥99.0% ACS, Fluka™
Potassium dihydrogen phosphate ≥99.0% ACS, Fluka™
Certificates
Potassium dihydrogen phosphate ≥99.0% ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Loss on drying (105°C, 2 h):
    Max. 0.2 %
  • pH (5 %, 25 °C):
    4.1 - 4.5
  • Water insoluble matter:
    Max. 0.01 %
  • Iron (Fe):
    Max. 0.002 %
  • Sodium (Na):
    Max. 0.005 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.003 %
  • pKa (25 °C):
    (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

Specification Test Results

Related Information

Frequently Bought Together