Order Entry
Austria
ContactUsLinkComponent
 
Potassium dihydrogen phosphate, anhydrous 99.5-100.5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. buffer substance, Fluka™
 
Potassium dihydrogen phosphate, anhydrous 99.5-100.5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. buffer substance, Fluka™
 

 

  • Assay (alk., calculated to dried substances):
    99.5 - 100.5 %
  • Assay:
    Min. 99.0 %
  • Loss on drying (130°C):
    Max. 0.2 %
  • Water insoluble matter:
    Max. 0.005 %
  • pH (5 %, 25 °C):
    4.2 - 4.5
  • Arsenic (As):
    Max. 0.5 ppm
  • Calcium (Ca):
    Max. 0.001 %
  • Cadmium (Cd):
    Max. 0.0005 %
  • Cobalt (Co):
    Max. 0.0005 %
  • Chromium (Cr):
    Max. 0.0015 %
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 0.0005 %
  • Manganese (Mn):
    Max. 0.0005 %
  • Nickel (Ni):
    Max. 0.0005 %
  • Sodium (Na):
    Max. 50 ppm
  • Lead (Pb):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 30 ppm
  • Total N:
    Max. 0.001 %
  • KMnO₄ reducing matter (as O):
    complying
  • Appearance of the solution:
    complying
  • Appearance (color):
    complying
  • Appearance (form):
    complying

 

 

Frequently Bought Together