Dipotassium hydrogen orthophosphate trihydrate ≥99.0% (by titrimetric analysis), Puriss. p.a., Fluka™
HONC60349-25KG
:
- Pack type:Fiberboard box
- Pk:25 kg
- Seq:4283
- Assay:99.0 - 101.0 %
- pH (5 %, 25 °C):9.1 - 9.4
- Arsenic (As):Max. 0.1 ppm
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 500 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Chloride (Cl):Max. 20 ppm
- Total S (as SO₄):Max. 50 ppm
- Total nitrogen (as N):Max. 10 ppm
- Solution 20g + 80 mL (H₂O):complying
:
:228.22 g/mol
:∼340 °C (1013 hPa)
:2.504 g/cm³ (20 °C)
:Ambient
:231-834-5
:16788-57-1
:7778-77-0
:K₂HPO₄·3H₂O
:MFCD00149926