

Potassium hydroxide ∼45% in aqueous solution for HPLC, Fluka™
:1310-58-3
:
:KOH
:MFCD00003553
:56.11 g/mol
:112 °C (1.013 hPa)
:–6 °C
:1.510 g/cm³ (20 °C)
:Ambient
:1814
:8,II
:GHS05
:GHS07
- Assay:40.5 - 49.5 %
- Assay of K₂CO₃:Max. 0.3 %
- Suitability test (HPLC):complying
- Density (D 20/4):1.440 - 1.480
- Absorbance at 500 nm:Max. 0.02
- Absorbance at 310 nm:Max. 0.02
- Absorbance at 300 nm:Max. 0.03
- Absorbance at 290 nm:Max. 0.05
- Absorbance at 240 nm:Max. 0.1
Frequently Bought Together