Order Entry
Austria
ContactUsLinkComponent
 
 
 
Potassium hydroxide 85-100.5%, pellets, Puriss., meets analytical specification of BP, Ph. Eur., NF, Fluka™
 
 
 
Potassium hydroxide 85-100.5%, pellets, Puriss., meets analytical specification of BP, Ph. Eur., NF, Fluka™
 

 

  • Assay:
    85.0 - 100.5 %
  • Total alkali (as KOH):
    Min. 85.0 %
  • Assay of K₂CO₃:
    Max. 1.0 %
  • Assay of Potassium (K):
    Min. 59.9 %
  • Aluminium (Al):
    Max. 10 ppm
  • Arsenic (As):
    Max. 0.0003 %
  • Calcium (Ca):
    Max. 0.002 %
  • Iron (Fe):
    Max. 10 ppm
  • Sodium (Na):
    Max. 1.0 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 100 ppm
  • Phosphate (PO₄):
    Max. 10 ppm
  • Silicate (as SiO₂):
    Max. 0.01 %
  • Sulfate (SO₄):
    Max. 100 ppm
  • Total N:
    Max. 0.0005 %
  • Appearance of the solution:
    complying
  • Water insoluble substances:
    complying
  • Residual solvents:
    complying

 

 

Frequently Bought Together