Order Entry
Austria
ContactUsLinkComponent
tert-Butyl methyl ether ≥99.0% ACS, Riedel-de Haën™
tert-Butyl methyl ether ≥99.0% ACS, Riedel-de Haën™
Catalog # HONC443808-20L
CAS Number:  
tert-Butyl methyl ether ≥99.0% ACS, Riedel-de Haën™
Catalog # HONC443808-20L
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    20 L
  • Seq:
    5302
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (GC):
    min. 99.0 %
  • Non-volatile matter:
    max. 0.001 %
  • Water (Karl Fischer):
    max. 0.05 %
  • Peroxides (as H2O2):
    max. 1 ppm
  • APHA:
    max. 10

Specification Test Results

About this item

Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether
 :
 :88.15 g/mol
Merck Index:13,06059
Boiling Pt:55.3 °C
Melting Pt:-109 °C
Melting Pt:–109 °C
Density:0.74 g/cm3
Flash Pt:-28 °C
Flash Pt:–28 °C
Storage Temperature:Ambient
EINECS:216-653-1
UN:2398
ADR:3,II
 :GHS07
 :GHS02
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812