Order Entry
Austria
ContactUsLinkComponent
 
Dimethyl sulphoxide ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
    
 
undefined
Dimethyl sulphoxide ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

 

 

  • Assay (GC):
    Min. 99.9 %
  • Refractive index (n 20/D):
    1.477 - 1.480
  • Water (Karl Fischer):
    Max. 0.04 %
  • UV-transmittance at 268 nm:
    Min. 30 %
  • UV-transmittance at 275 nm:
    Min. 60 %
  • UV-transmittance at 300 nm:
    Min. 85 %
  • UV-transmittance at 350 nm:
    Min. 95 %
  • UV-transmittance from 400 nm:
    Min. 98 %
  • Suitability for the GC Headspace:
    complying
  • Vapor Pressure:
    0.42 mmHg ( 20 °C)
  • Explosion Limit:
    42 %, 63 °F

 

 

Frequently Bought Together