Order Entry
Austria
ContactUsLinkComponent
Ammonium cerium(IV) nitrate 0.1 M Ce(NH4)2(NO3)6 in aqueous solution ≥97% Reag. Ph. Eur. volumetric solution, Fluka™
 
 
Ammonium cerium(IV) nitrate 0.1 M Ce(NH4)2(NO3)6 in aqueous solution ≥97% Reag. Ph. Eur. volumetric solution, Fluka™
  38187-1L
 :  
 
 
Ammonium cerium(IV) nitrate 0.1 M Ce(NH4)2(NO3)6 in aqueous solution ≥97% Reag. Ph. Eur. volumetric solution, Fluka™
  38187-1L
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Appearance:
    complying
  • Assay:
    0.095 - 0.105 mol/l
  • Density (D 20/4):
    1.083 - 1.098

 

 

 :
 :548.22 g/mol
 :1.1 g/cm³
 :Ambient
 :2796
 :8,II
 :GHS05
 :GHS07
 :16774-21-3
 :(NH₄)₂Ce(NO₃)₆
 :MFCD00151121