Order Entry
Austria
ContactUsLinkComponent
 
Sodium acetate trihydrate 99.0-101.0% (dried basis), Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E262 (max. 0.00002% Al), Fluka™
 
undefined
Sodium acetate trihydrate 99.0-101.0% (dried basis), Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E262 (max. 0.00002% Al), Fluka™

 

  • Assay (calculated to the dried substance):
    99.0 - 101.0 %
  • Loss on drying (130°C):
    39.0 - 40.5 %
  • pH (5 %, 25 °C):
    7.5 - 9.0
  • Water insoluble matter:
    Max. 0.05 %
  • Free alkali (as Na₂CO₃):
    Max. 0.05 %
  • Aluminium (Al):
    Max. 0.2 ppm
  • Arsenic (As):
    Max. 2 ppm
  • Calcium (Ca):
    Max. 20 ppm
  • Calcium, Magnesium:
    complying
  • Copper (Cu):
    Max. 0.001 %
  • Iron (Fe):
    Max. 10 ppm
  • Mercury (Hg):
    Max. 1 ppm
  • Potassium (K):
    Max. 0.02 %
  • Magnesium (Mg):
    Max. 20 ppm
  • Lead (Pb):
    Max. 2 ppm
  • Zinc (Zn):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Chloride (Cl):
    Max. 50 ppm
  • Sulfate (SO₄):
    Max. 50 ppm
  • KMnO₄ reducing matter (as HCOOH):
    Max. 0.1 %
  • KMnO₄ reducing matter (as O):
    Max. 0.002 %
  • Formic acid, formiates other impurities (HCOOH):
    Max. 1000 ppm
  • Appearance of the solution:
    complying
  • Reducing impurities:
    complying
  • Residual solvents:
    complying

 

 

Frequently Bought Together