Ammonium carbonate, Puriss., meets analytical specification of NF, Ph. Franc., FCC, Fluka™
:506-87-6
:10361-29-2
:(NH₄)₂CO₃
:MFCD00010890
:96.09 g/mol
:13,00508
:>59 °C
:0.9 g/cm³ (20 °C)
:208-058-0
:GHS07
:Ambient
- Vapor Density:2.7 (vs air)
- Assay of NH₃:30.0 - 34.0 %
- Non-volatile matter:Max. 0.05 %
- Residue on ignition (SO₄):Max. 0.05 %
- Arsenic (As):Max. 0.0003 %
- Iron (Fe):Max. 0.0005 %
- Lead (Pb):Max. 0.0003 %
- Heavy metals (as Pb):Max. 0.0005 %
- Chloride (Cl):Max. 0.001 %
- Sulfate (SO₄):Max. 0.005 %
- Appearance of the solution:complying
- Residual solvents:complying
Frequently Bought Together