
Orthoboric acid 99.5-100.5%, powder, Puriss., meets analytical specification of BP, Ph. Eur., NF, Fluka™
:10043-35-3
:
:B(OH)₃
:MFCD00011337
:13,01326
:171 °C
:1.510 g/cm³ (20 °C)
:61.83 g/mol
:233-139-2
:01-2119486683-25
:GHS08
:Ambient
- Assay:99.5 - 100.5 %
- Loss on drying (over silicagel, 5h):Max. 0.5 %
- pH (3,3 %, 20 °C):3.8 - 4.8
- Arsenic (As):Max. 0.0005 %
- Iron (Fe):Max. 0.0005 %
- Heavy metals (as Pb):Max. 0.001 %
- Sulfate (SO₄):Max. 0.04 %
- Organic matter:complying
- Residual solvents:complying
- Solubility in C₂H₅OH:complying
- Appearance of the solution:complying
- Completeness of the solution:complying
Frequently Bought Together