
Sulphuric acid ≥93%, TraceSELECT™ for trace analysis, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Merck Index:14,08974
Boiling Pt:310 °C (1.013 hPa)
Melting Pt:–14…–10 °C
Density:1.840 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05
Specification Test Results
- Assay:93.0 - 98.0 %
- Silver (Ag):Max. 1 ppb
- Aluminium (Al):Max. 5 ppb
- Arsenic (As):Max. 0.001 ppm
- Gold (Au):Max. 1 ppb
- Barium (Ba):Max. 1 ppb
- Beryllium (Be):Max. 0.5 ppb
- Bismuth (Bi):Max. 0.5 ppb
- Calcium (Ca):Max. 20 ppb
- Cadmium (Cd):Max. 0.5 ppb
- Cerium (Ce):Max. 0.5 ppb
- Cobalt (Co):Max. 0.5 ppb
- Chromium (Cr):Max. 1 ppb
- Cesium (Cs):Max. 0.5 ppb
- Copper (Cu):Max. 1 ppb
- Iron (Fe):Max. 10 ppb
- Gallium (Ga):Max. 0.5 ppb
- Germanium (Ge):Max. 1 ppb
- Mercury (Hg):Max. 0.5 ppb
- Potassium (K):Max. 10 ppb
- Lithium (Li):Max. 0.5 ppb
- Magnesium (Mg):Max. 5 ppb
- Manganese (Mn):Max. 0.5 ppb
- Molybdenum (Mo):Max. 0.5 ppb
- Sodium (Na):Max. 20 ppb
- Nickel (Ni):Max. 2 ppb
- Lead (Pb):Max. 1 ppb
- Palladium (Pd):Max. 0.5 ppb
- Platinum (Pt):Max. 0.5 ppb
- Rubidium (Rb):Max. 0.5 ppb
- Rhodium (Rh):Max. 0.5 ppb
- Antimony (Sb):Max. 1 ppb
- Selenium (Se):Max. 0.005 ppm
- Tin (Sn):Max. 1 ppb
- Strontium (Sr):Max. 0.5 ppb
- Titanium (Ti):Max. 1 ppb
- Thallium (Tl):Max. 0.5 ppb
- Vanadium (V):Max. 1 ppb
- Zinc (Zn):Max. 5 ppb
- Zirconium (Zr):Max. 0.5 ppb
- Chloride (Cl):Max. 0.1 ppm
- Nitrate (NO₃):Max. 0.2 ppm
- Phosphate (PO₄):Max. 0.5 ppm
- Vapor Density:<0.3 (25 °C, vs air)
Specification Test Results
Frequently Bought Together