Order Entry
Austria
ContactUsLinkComponent
 
 
Sulphuric acid 95.0-97.0%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. for the determination of mercury, Fluka™
 
 
Sulphuric acid 95.0-97.0%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. for the determination of mercury, Fluka™
 

 

  • Vapor Pressure:
    1 mmHg ( 146 °C)
  • Assay:
    95.0 - 97.0 %
  • Residue on ignition:
    Max. 5 ppm
  • Ammonium (NH₄):
    Max. 2 ppm
  • Silver (Ag):
    Max. 0.000002 %
  • Aluminium (Al):
    Max. 0.000005 %
  • Arsenic (As):
    Max. 0.01 ppm
  • Barium (Ba):
    Max. 0.000005 %
  • Beryllium (Be):
    Max. 0.000001 %
  • Bismuth (Bi):
    Max. 0.000005 %
  • Calcium (Ca):
    Max. 0.00002 %
  • Cadmium (Cd):
    Max. 0.000002 %
  • Cobalt (Co):
    Max. 0.000001 %
  • Chromium (Cr):
    Max. 0.000005 %
  • Copper (Cu):
    Max. 0.000001 %
  • Iron (Fe):
    Max. 0.1 ppm
  • Germanium (Ge):
    Max. 0.000005 %
  • Mercury (Hg):
    Max. 0.005 ppm
  • Potassium (K):
    Max. 0.00001 %
  • Lithium (Li):
    Max. 0.000001 %
  • Magnesium (Mg):
    Max. 0.000005 %
  • Manganese (Mn):
    Max. 0.000001 %
  • Molybdenum (Mo):
    Max. 0.000002 %
  • Sodium (Na):
    Max. 0.00005 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.000002 %
  • Strontium (Sr):
    Max. 0.000002 %
  • Titanium (Ti):
    Max. 0.00001 %
  • Thallium (Tl):
    Max. 0.000005 %
  • Vanadium (V):
    Max. 0.000001 %
  • Zinc (Zn):
    Max. 0.000005 %
  • Zirconium (Zr):
    Max. 0.00001 %
  • Chloride (Cl):
    Max. 0.1 ppm
  • Nitrate (NO₃):
    Max. 0.2 ppm
  • Phosphate (PO₄):
    Max. 0.00005 %
  • KMnO₄ reducing matter (as O):
    Max. 0.0002 %
  • APHA:
    Max. 10
  • KMnO₄ reducing matter (as SO₂):
    Max. 2 ppm

 

 

Frequently Bought Together