


Nitric acid ≥65%, Puriss. p.a. ISO, Reag. Ph. Eur. for determinations with dithizone, Fluka™
:7697-37-2
:
:HNO₃
:MFCD00011349
:63.01 g/mol
:13,06608
:122 °C (1.013 hPa)
:–31 °C
:1.400…1.480 g/cm³ (20 °C)
:Ambient
:231-714-2
:2031
:8,II
:GHS06
:GHS05
:GHS03
- Appearance:complying
- Assay:65.0 - 67.0 %
- Density (D 20/4):1.390 - 1.410
- Residue on ignition (SO₄):Max. 0.0005 %
- Silver (Ag):Max. 0.000001 %
- Arsenic (As):Max. 0.000001 %
- Aluminium (Al):Max. 0.000005 %
- Barium (Ba):Max. 0.000001 %
- Beryllium (Be):Max. 0.000001 %
- Bismuth (Bi):Max. 0.00001 %
- Calcium (Ca):Max. 0.00001 %
- Cadmium (Cd):Max. 0.000001 %
- Cobalt (Co):Max. 0.000001 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000001 %
- Iron (Fe):Max. 0.00001 %
- Germanium (Ge):Max. 0.000005 %
- Mercury (Hg):Max. 0.000001 %
- Potassium (K):Max. 0.000005 %
- Lithium (Li):Max. 0.000001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000001 %
- Molybdenum (Mo):Max. 0.000002 %
- Sodium (Na):Max. 0.00005 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.000001 %
- Strontium (Sr):Max. 0.000001 %
- Titanium (Ti):Max. 0.00001 %
- Thallium (Tl):Max. 0.000005 %
- Vanadium (V):Max. 0.000001 %
- Zinc (Zn):Max. 0.000005 %
- Zirconium (Zr):Max. 0.00001 %
- Heavy metals (as Pb):Max. 0.0002 %
- Chloride (Cl):Max. 0.00005 %
- Phosphate (PO₄):Max. 0.00005 %
- Sulfate (SO₄):Max. 0.00005 %
- Suitability for determinations with dithizone:complying
- Residual solvents:complying
Frequently Bought Together