Order Entry
Austria
ContactUsLinkComponent
Potassium nitrate ≥99.995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Potassium nitrate ≥99.995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Catalog # 60429-100G
CAS Number:  
Potassium nitrate ≥99.995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Catalog # 60429-100G
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :101.1 g/mol
Merck Index:13,07733
Melting Pt:334 °C
ADR:5.1,III
REACH:01-2119488224-35
CAS Number:7757-79-1
Formula:KNO₃
MDL Number:MFCD00011409