Order Entry
Austria
ContactUsLinkComponent
 
 
Potassium nitrate 99.0-100.5%, Puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E252, Fluka™
 
 
Potassium nitrate 99.0-100.5%, Puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E252, Fluka™

 

  • Assay:
    99.0 - 100.5 %
  • Assay (calculated to the dried substance):
    99.0 - 100.5 %
  • Loss on drying (105°C):
    Max. 0.5 %
  • pH (5 %, 20°C):
    5-8
  • Ammonium (NH₄):
    Max. 50 ppm
  • Arsenic (As):
    Max. 1 ppm
  • Calcium (Ca):
    Max. 50 ppm
  • Copper (Cu):
    Max. 10 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Mercury (Hg):
    Max. 1 ppm
  • Sodium (Na):
    Max. 0.1 %
  • Lead (Pb):
    Max. 2 ppm
  • Zinc (Zn):
    Max. 10 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Chloride (Cl):
    Max. 10 ppm
  • Nitrite (NO₂):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 100 ppm
  • Chlorate (ClO₃):
    complying
  • Appearance of the solution:
    complying
  • Acidity or alkalinity:
    complying
  • Reducible impurities:
    complying
  • Residual solvents:
    complying
  • Identity on K:
    complying
  • Identity on NO₃:
    complying

 

 

Frequently Bought Together