Order Entry
Austria
ContactUsLinkComponent
Potassium permanganate ≥99.0% ACS, Fluka™
Irritant
Corrosive
Oxidizing
Potassium permanganate ≥99.0% ACS, Fluka™
Certificates
Irritant
Corrosive
Oxidizing
Potassium permanganate ≥99.0% ACS, Fluka™

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Water insoluble matter:
    Max. 0.2 %
  • Chloride, Chlorate (as Cl):
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 0.02 %

Specification Test Results

Related Information

Frequently Bought Together