Sodium chloride ≥99.5% for HPLC, Fluka™
Catalog # 73575-250G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.5 - 100.5 %
- pH (10 %, 20°C):5.0 - 8.0
- Absorbance at 220 nm:Max. 0.05
- Absorbance at 230 nm:Max. 0.03
- Absorbance at 240 nm:Max. 0.02
- Absorbance at 260 nm:Max. 0.01
- Absorbance 500 nm:Max. 0.01
- Appearance of the solution (c=10%):complying
Specification Test Results
About this item
Synonyms: Saline, NaCl, Sea salt, Halite
:
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1.465 °C (1.013 hPa)
Melting Pt:800 °C
Density:2.160 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477