Order Entry
Austria
ContactUsLinkComponent
n-Dodecane ≥99%, Reagent Grade
Health
n-Dodecane ≥99%, Reagent Grade
Certificates
undefined
Health
n-Dodecane ≥99%, Reagent Grade

Specification Test Results

  • Vapor Pressure:
    1 mmHg ( 47.8 °C)
  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99 %
  • Refractive index (n 20/D):
    1.420 - 1.424
  • APHA:
    Max. 20
  • Vapor Density:
    5.96 (vs air)
  • Autoignition Temperature:
    401 °F

Specification Test Results

Related Information

Frequently Bought Together