Order Entry
Austria
ContactUsLinkComponent
Ethylene glycol ≥99.5%, Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Health
Ethylene glycol ≥99.5%, Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Certificates
undefined
Health
Ethylene glycol ≥99.5%, Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Certificates

Specification Test Results

  • Refractive Index:
    n20/D 1.431 (lit.)
  • Assay (GC):
    Min. 99.5 %
  • Identity (IR):
    complying
  • Boiling range:
    195 - 197 °C
  • Density (D 20/20):
    1.113 - 1.115
  • Refractive index (n 20/D):
    1.430 - 1.432
  • Water (Karl Fischer):
    Max. 0.1 %
  • Sulfated ash:
    Max. 0.005 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.2 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.05 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 1 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.2 ppm
  • Chloride (Cl):
    Max. 0.2 ppm
  • Sulfate (SO₄):
    Max. 10 ppm
  • Formaldehyde:
    Max. 50 ppm
  • KMnO₄ reducing matter (as O):
    Max. 0.0003 %
  • APHA:
    Max. 25
  • Explosion Limit:
    15.3 %
  • Ignition Residue:
    ≤0.005% (as SO₄)

Specification Test Results

Related Information

Frequently Bought Together