

2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:77…83 °C (1.013 hPa)
Melting Pt:–90 °C
Density:0.784…0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Vapor Pressure:44 mmHg ( 25 °C)
- Assay (GC):Min. 99.8 %
- Boiling range:81 - 83 °C
- Density (D 20/4):0.784 - 0.786
- Refractive index (n 20/D):1.3770 - 1.3780
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.1 %
- Free acid (as C₂H₅COOH):Max. 0.00074 %
- Free alkali (as NH₃):Max. 0.00017 %
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Bismuth (Bi):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Potassium (K):Max. 0.00005 %
- Lithium (Li):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Molybdenum (Mo):Max. 0.00001 %
- Sodium (Na):Max. 0.0001 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Strontium (Sr):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- APHA:Max. 10
- Carbonyl compounds (as CO):Max. 0.005 %
- Carbonyl compounds (ACS):complying
- Ethanol (GC):Max. 0.01 %
- Methanol (GC):Max. 0.1 %
- KMnO₄ reducing matter (as O):Max. 0.0005 %
- Solubility in water:complying
- Peroxides (as H₂O₂):Max. 0.00034 %
- Reaction against H₂SO₄:complying
Specification Test Results
Frequently Bought Together