

2-Propanol ≥99.5% ACS, Riedel-de Haën™
:67-63-0
:
:(CH₃)₂CHOH
:MFCD00011674
:60.1 g/mol
:77…83 °C (1.013 hPa)
:–90 °C
:0.784…0.786 g/cm³ (20 °C)
:12 °C
:Ambient
:200-661-7
:1219
:3,II
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Explosion Limit:2 %
- Assay (GC):Min. 99.50 %
- IR spectrum:complying
- Solubility in water:complying
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.2 %
- Free acid, free alkali:Max. 0.0001 meq/g
- Carbonyl compounds (as Acetone):Max. 0.002 %
- Carbonyl compounds (as propionaldehyde):Max. 0.002 %
- APHA:Max. 10
- Autoignition Temperature:750 °F
- Refractive Index:n20/D 1.377 (lit.)
- Vapor Pressure:33 mmHg ( 20 °C)
Frequently Bought Together