Order Entry
Austria
ContactUsLinkComponent
1-Propanol ≥99% (by GC), Purum
1-Propanol ≥99% (by GC), Purum
Certificates
undefined
1-Propanol ≥99% (by GC), Purum

Specification Test Results

  • Assay (GC):
    Min. 99 %
  • Non-volatile matter:
    Max. 0.005 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as C₂H₅COOH):
    Max. 0.002 %
  • Autoignition Temperature:
    700 °F
  • Vapor Pressure:
    10 mmHg ( 147 °C)
  • Vapor Density:
    2.1 (vs air)
  • Refractive Index:
    n20/D 1.384 (lit.)

Specification Test Results

Related Information

Frequently Bought Together