Order Entry
Austria
ContactUsLinkComponent
 
 
 
Acetone ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
    
 
undefined
 
 
Acetone ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

 

 

  • Assay (GC):
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as CH₃COOH):
    Max. 0.002 %
  • Transmittance at 330 nm:
    Min. 15 %
  • Transmittance at 335 nm:
    Min. 50 %
  • Transmittance at 340 nm:
    Min. 80 %
  • Transmittance from 350 nm:
    Min. 98 %
  • Vapor Density:
    2 (vs air)
  • Vapor Pressure:
    184 mmHg ( 20 °C)

 

 

Frequently Bought Together