Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
    
 
undefined
 
 
 
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Assay (GC):
    Min. 99.9 %
  • Propionitrile (GC):
    Max. 0.2 %
  • Non-volatile matter:
    Max. 0.0002 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Free alkali (as NH₃):
    Max. 0.0001 %
  • Silver (Ag):
    Max. 0.1 ppm
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.05 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.02 ppm
  • Potassium (K):
    Max. 0.05 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.02 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 0.05 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.02 ppm
  • Tin (Sn):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.1 ppm
  • HPLC Gradient at 210 nm:
    Max. 1 mAU
  • HPLC Gradient at 254 nm:
    Max. 0.2 mAU
  • Baseline drift at 210 nm:
    Max. 12 mAU
  • Suitability for the LC-MS:
    complying
  • Fluorescence (chinin) at 254 nm:
    Max. 0.5 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 0.5 ppb
  • Absorbance at 195 nm:
    Max. 0.097
  • Absorbance at 200 nm:
    Max. 0.022
  • Absorbance at 230 nm:
    Max. 0.004
  • Refractive Index:
    n20/D 1.344 (lit.)

 

 

Frequently Bought Together