Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for HPLC, for UV spectrophotometry, Riedel-de Haën™
    
 
undefined
 
 
 
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for HPLC, for UV spectrophotometry, Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

 

 

  • Assay (GC):
    Min. 99.9 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Free acid (as CH₃COOH):
    Max. 0.002 %
  • Free alkali (as NH₃):
    Max. 0.0002 %
  • Transmittance at 195 nm:
    Min. 65 %
  • Transmittance at 200 nm:
    Min. 88 %
  • Transmittance at 210 nm:
    Min. 93 %
  • Transmittance at 235 nm:
    Min. 99 %
  • Absorbance (JP)(Val.Ref):
    complying
  • Fluorescence (chinin) at 254 nm:
    Max. 1 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 0.5 ppb

 

 

Frequently Bought Together