Order Entry
Austria
ContactUsLinkComponent
Acetonitrile ≥99.8% (by GC), CHROMASOLV™ for liquid chromatography, Riedel-de Haën™
Health
Irritant
Flammable
Acetonitrile ≥99.8% (by GC), CHROMASOLV™ for liquid chromatography, Riedel-de Haën™
   Sustainable Options Available
Certificates
undefined
Health
Irritant
Flammable
Acetonitrile ≥99.8% (by GC), CHROMASOLV™ for liquid chromatography, Riedel-de Haën™
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Refractive Index:
    n20/D 1.344 (lit.)
  • Assay (GC):
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Transmittance at 200 nm:
    Min. 50 %
  • Transmittance at 220 nm:
    Min. 90 %
  • Transmittance from 240 nm:
    Min. 99 %
  • Vapor Pressure:
    72.8 mmHg ( 20 °C)
  • Explosion Limit:
    16 %
  • Autoignition Temperature:
    973 °F

Specification Test Results

Related Information

Frequently Bought Together