Order Entry
Austria
ContactUsLinkComponent
 
Calcium chloride dihydrate ≥99% ACS, Fluka™
  223506-4X2.5KG
 :  
Calcium chloride dihydrate ≥99% ACS, Fluka™
  223506-4X2.5KG
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    10 kg
  • Seq:
    4082
  • Pk Info:
    4 x 2.5 kg

 

 

  • Assay:
    99.0 - 105.0 %
  • pH (5 %, 25 °C):
    4.5 - 8.5
  • Water insoluble matter:
    Max. 0.01 %
  • Barium (Ba):
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.01 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Sodium (Na):
    Max. 0.02 %
  • Strontium (Sr):
    Max. 0.1 %
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 0.01 %
  • Ammonium (NH₄):
    Max. 0.005 %
  • Oxidizing substances (as NO₃):
    Max. 0.003 %

 

 

 :
 :147.01 g/mol
 :13,01660
 :1.900 °C (1.013 hPa)
 :765 °C
 :GHS07
 :Ambient
 :10035-04-8
 :CaCl₂·2H₂O
 :MFCD00149613