Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate dihydrate ≥98.5% for HPLC, Fluka™
Disodium hydrogen phosphate dihydrate ≥98.5% for HPLC, Fluka™
Catalog # 71633-250G
CAS Number:  
Disodium hydrogen phosphate dihydrate ≥98.5% for HPLC, Fluka™
Catalog # 71633-250G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    98.5 - 102.0 %
  • pH (10 %):
    8.8 - 9.4
  • Colour of the solution:
    complying
  • Turbidity of solution:
    complying
  • Absorbance 500 nm:
    Max. 0.02
  • Absorbance at 320 nm:
    Max. 0.02
  • Absorbance 280 nm:
    Max. 0.04
  • Absorbance at 260 nm:
    Max. 0.06
  • Absorbance at 230 nm:
    Max. 0.1

Specification Test Results

About this item

 :
 :177.99 g/mol
Melting Pt:450 °C
Density:2.07 g/cm³ (16 °C)
Storage Temperature:Ambient
CAS Number:10028-24-7
Formula:Na₂HPO₄·2H₂O
MDL Number:MFCD00149182