Single - component oxygenates solutions
Supplier: Restek
About this item
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
10 Options Available Below
Product Details & Documents
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- REST30466
- REST30467
- REST30402
- REST30627
- REST30628
- REST30629
- REST30617
- REST30631
- REST30618
- REST30470
- Description:
- Ethanol, with 10000 µg/ml in DI water
- Methanol, with 10000 µg/ml in DI water
- Methyl tert-butyl ether (MTBE), with 2000 µg/ml in purge and trap grade methanol
- Diisopropyl ether (DIPE), with 2000 µg/ml in purge and trap grade methanol
- Ethyl tert-butyl ether (ETBE), with 2000 µg/ml in purge and trap grade methanol
- tert-Amyl methyl ether (TAME), with 2000 µg/ml in purge and trap grade methanol
- tert-Amyl ethyl ether (TAEE), with 2000 µg/ml in purge and trap grade methanol
- tert-Amyl alcohol, with 10000 µg/ml in purge and trap grade methanol
- tert-Butanol-d9, with 20000 µg/ml in purge and trap grade methanol
- tert-Butanol, with 50000 µg/ml in purge and trap grade methanol
- CAS No.:
- 64-17-5
- 67-56-1
- 1634-04-4
- 108-20-3
- 637-92-3
- 994-05-8
- 919-94-8
- 25725-11-5
- 75-65-0
- Formula:
- C₂H₅OH
- CH₃OH
- (CH₃)₃COCH₃
- C₆H₁₄O
- C₆H₁₄O
- C₆H₁₄O
- C₇H₁₆O
- (CD₃)₃COH
- (CH₃)₃COH
Specifications
Product Family Options
Product Information
- StandardsPk InfoPkAvailabilityPrice
- REST30466Ethanol, with 10000 µg/ml in DI waterEthanol, with 10000 µg/ml in DI waterUS EPA 502.2, 8021, 8206B1 ml/ampoule1Log in for availability1 * 1 ST56,50 €Specifications:
More
Specifications
Cat. No.REST30466DescriptionEthanol, with 10000 µg/ml in DI waterCAS No.64-17-5FormulaC₂H₅OH - REST30467Methanol, with 10000 µg/ml in DI waterMethanol, with 10000 µg/ml in DI waterUS EPA 502.2, 8021, 8206B1 ml/ampoule1 mLLog in for availability1 * 1 mL54,50 €Specifications:
More
Specifications
Cat. No.REST30467DescriptionMethanol, with 10000 µg/ml in DI waterCAS No.67-56-1FormulaCH₃OH - REST30402Methyl tert-butyl ether (MTBE), with 2000 µg/ml in purge and trap grade methanolMethyl tert-butyl ether (MTBE), with 2000 µg/ml in purge and trap grade methanolUS EPA 502.2, 8021, 8206B1 ml/ampoule1 mLLog in for availability1 * 1 mL52,50 €Specifications:
More
Specifications
Cat. No.REST30402DescriptionMethyl tert-butyl ether (MTBE), with 2000 µg/ml in purge and trap grade methanolCAS No.1634-04-4Formula(CH₃)₃COCH₃ - REST30627Diisopropyl ether (DIPE), with 2000 µg/ml in purge and trap grade methanolDiisopropyl ether (DIPE), with 2000 µg/ml in purge and trap grade methanolUS EPA 502.2, 8021, 8206B1 ml/ampoule1 AmpoulLog in for availability1 * 1 Ampull41,00 €Specifications:
More
Specifications
Cat. No.REST30627DescriptionDiisopropyl ether (DIPE), with 2000 µg/ml in purge and trap grade methanolCAS No.108-20-3FormulaC₆H₁₄O - REST30628Ethyl tert-butyl ether (ETBE), with 2000 µg/ml in purge and trap grade methanolEthyl tert-butyl ether (ETBE), with 2000 µg/ml in purge and trap grade methanolUS EPA 502.2, 8021, 8206B1 ml/ampoule1 AmpoulLog in for availability1 * 1 Ampull52,50 €Specifications:
More
Specifications
Cat. No.REST30628DescriptionEthyl tert-butyl ether (ETBE), with 2000 µg/ml in purge and trap grade methanolCAS No.637-92-3FormulaC₆H₁₄O - REST30629tert-Amyl methyl ether (TAME), with 2000 µg/ml in purge and trap grade methanoltert-Amyl methyl ether (TAME), with 2000 µg/ml in purge and trap grade methanolUS EPA 502.2, 8021, 8206B1 ml/ampoule1 AmpoulLog in for availability1 * 1 Ampull53,00 €Specifications:
More
Specifications
Cat. No.REST30629Descriptiontert-Amyl methyl ether (TAME), with 2000 µg/ml in purge and trap grade methanolCAS No.994-05-8FormulaC₆H₁₄O - REST30617tert-Amyl ethyl ether (TAEE), with 2000 µg/ml in purge and trap grade methanoltert-Amyl ethyl ether (TAEE), with 2000 µg/ml in purge and trap grade methanolUS EPA 502.2, 8021, 8206B1 ml/ampoule1 AmpoulLog in for availability1 * 1 Ampull46,00 €Specifications:
More
Specifications
Cat. No.REST30617Descriptiontert-Amyl ethyl ether (TAEE), with 2000 µg/ml in purge and trap grade methanolCAS No.919-94-8FormulaC₇H₁₆O - REST30631tert-Amyl alcohol, with 10000 µg/ml in purge and trap grade methanoltert-Amyl alcohol, with 10000 µg/ml in purge and trap grade methanolUS EPA 502.2, 8021, 8206B1 ml/ampoule1 AmpoulLog in for availability1 * 1 Ampull46,00 €Specifications:
More
Specifications
Cat. No.REST30631Descriptiontert-Amyl alcohol, with 10000 µg/ml in purge and trap grade methanolCAS No.Formula - REST30618tert-Butanol-d9, with 20000 µg/ml in purge and trap grade methanoltert-Butanol-d9, with 20000 µg/ml in purge and trap grade methanolUS EPA 502.2, 8021, 8206B1 ml/ampoule1 AmpoulLog in for availability1 * 1 AmpullSpecifications:
More
Specifications
Cat. No.REST30618Descriptiontert-Butanol-d9, with 20000 µg/ml in purge and trap grade methanolCAS No.25725-11-5Formula(CD₃)₃COH - REST30470tert-Butanol, with 50000 µg/ml in purge and trap grade methanoltert-Butanol, with 50000 µg/ml in purge and trap grade methanolUS EPA 502.2, 8021, 8206B1 ml/ampoule1 mLLog in for availability1 * 1 mL54,50 €Specifications:
More
Specifications
Cat. No.REST30470Descriptiontert-Butanol, with 50000 µg/ml in purge and trap grade methanolCAS No.75-65-0Formula(CH₃)₃COH



