Unleaded gasolines (ASTM Class 2 Accelerant)
: Restek
The weathered materials indicate the percent weight loss from the original material. Prepared from a single source (one refinery) product.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Frequently Bought Together