About this item
These standards are used for pesticide residue quantification.
- Compatible with both LC-MS andGC-MS applications and for optimising LC-MS/MS system performance
- Would mitigate matrix effects
Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:163165-75-1
- Formula:C₈H₉D₅ClN₅
- Description:Atrazine-d5