Order Entry
Austria
ContactUsLinkComponent
Water with Formic acid 0.1% / Ammonium formate 2mM, HiPerSolv CHROMANORM® for LC-MS
Water with Formic acid 0.1% / Ammonium formate 2mM, HiPerSolv CHROMANORM® for LC-MS
Catalog # 85960.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Water with Formic acid 0.1% / Ammonium formate 2mM, HiPerSolv CHROMANORM® for LC-MS
Catalog # 85960.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Nominal concentration (V/V):
    0.095 to 0.105 %
  • Ammonium formate:
    2 mM
  • Ca (Calcium):
    Max. 0.2 ppm
  • K (Potassium):
    Max. 0.2 ppm
  • Mg (Magnesium):
    Max. 0.2 ppm
  • Na (Sodium):
    Max. 0.5 ppm
  • Transmittance (225 nm):
    Min. 20 %
  • Transmittance (235 nm):
    Min. 50 %
  • Transmittance (240 nm):
    Min. 80 %
  • Transmittance (250 nm):
    Min. 95 %
  • Transmittance (260 nm):
    Min. 99 %
  • Suitable for LC-MS:
    Passes test

Specification Test Results

About this item

 :