ContactUsLinkComponent
Preparative HPLC columns, Ultra Biphenyl
 :  Restek
undefined
Preparative HPLC columns, Ultra Biphenyl
 :  Restek
  • Catalog No:
  • REST9109518
  • REST9109519
  • REST9109577
  • Carbon load:
  • 15%
  • 15%
  • 15%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • pH range:
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • Surface area:
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • Temperature limit:
  • 80 °C
  • 80 °C
  • 80 °C
  • USP code:
  • L11
  • L11
  • L11
Frequently Bought Together