Manual microliter syringes, SGE
Supplier: Restek
Certificates
About this item
Each syringe plunger and barrel assembly is manufactured as one working unit; components are not interchangeable or individually replaceable.
- Available with fixed or removable needles
Frequently Bought Together