ASTM precision thermometers, with DAkkS calibration certificate
Supplier: Ludwig Schneider GmbH & CO. KG
Certificates
About this item
These precision thermometers are ideal to measure precisely and reliably without mercury from the very start.
- Round, white back capillary form
- Suitable for calibration
Blue filling stem type.
Certifications: Conform to ASTM standards.
Ordering information: Available with works certificate or with DAkkS calibration certificate (optional).
Packaging: Packed individually in a square plastic sleeve.
Frequently Bought Together