Iodine 0.025 M (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
85805.260
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
- Titer (20°C; real value 0.5 % accuracy):0,02488 --> 0,02512 mol/l
:
:253.81 g/mol
:GHS08
:Ambient
:7553-56-2
:I₂
:MFCD00011355