Iodine 0.0241 M (0.0482 N) in aqueous solution, AVS TITRINORM volumetric solution
85806.320
:
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:
- Titer (20°C; real value 0.5 % accuracy):0.02398 -> 0,02422 mol/l
- Content KI:57g/l +/-1 g
:
:253.81 g/mol
:GHS08
:Ambient
:7553-56-2
:I₂
:MFCD00011355