Order Entry
Austria
ContactUsLinkComponent
Mouse Recombinant SCF (Animal free) (from E. coli)
Mouse Recombinant SCF (Animal free) (from E. coli)
Certificates
undefined
Mouse Recombinant SCF (Animal free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
  • Gene ID:
    P20826
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    c-Kit ligand|steel factor|KL|MGF
  • Protein/peptide name:
    SCF
  • Purity:
    ≥95%
  • Molecular weight:
    18,4 kDa
  • Sequence:
    MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
  • Endotoxin level:
    Low
  • Formulation:
    20 mM sodium phosphate, pH 6,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together