Order Entry
Austria
ContactUsLinkComponent
Oxalic acid 0.005 mol/l (0.01 N), AVS TITRINORM volumetric solution
Oxalic acid 0.005 mol/l (0.01 N), AVS TITRINORM volumetric solution
Catalog # 89619.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Oxalic acid 0.005 mol/l (0.01 N), AVS TITRINORM volumetric solution
Catalog # 89619.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :90.04 g/mol
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573