Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide 1 mol/l (1 N) in methanol, AVS TITRINORM volumetric solution
Potassium hydroxide 1 mol/l (1 N) in methanol, AVS TITRINORM volumetric solution
Catalog # 89346.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium hydroxide 1 mol/l (1 N) in methanol, AVS TITRINORM volumetric solution
Catalog # 89346.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :56.11 g/mol
UN:2924
ADR:3 (8),II
 :GHS02
 :GHS06
 :GHS08
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553